Learn More
DYKDDDDK tag Mouse anti-Recombinant Protein, FITC, Clone: 2B3C4, Proteintech
Shop All Proteintech Group Inc Products

Description
The DYKDDDDK tag antibody from Proteintech is a mouse monoclonal antibody generated with peptide of DYKDDDDK tag. This antibody recognizes recombinant protein antigen. The DYKDDDDK tag antibody has been validated for the following applications: IF analysis. This antibody is fitc.
Specifications
Specifications
| Antigen | DYKDDDDK tag |
| Applications | Immunofluorescence |
| Classification | Monoclonal |
| Clone | 2B3C4 |
| Concentration | 1200 μg/mL |
| Conjugate | FITC |
| Host Species | Mouse |
| Quantity | 100 μL |
| Primary or Secondary | Primary |
| Isotype | IgG1 |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.