Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

DYKDDDDK Tag Mouse, Biotin, Clone: 5A8E5, THE™, GenScript™

Catalog No. 50273498
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-273-498 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-273-498 Supplier Genscript Corporation Supplier No. A01429100
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Reconstitute the lyophilized powder with deionized water (or equivalent) to an final concentration of 0.5 mg/mL. GenScript THETM DYKDDDDK Tag Antibody [Biotin], mAb, Mouse is the biotin conjugated THETM DYKDDDDK Tag Antibody, mAb, Mouse (A00187) purified from mice ascites by protein A affinity column. The antibody is suitable for detecting the expression level of DYKDDDDK in fusion proteins.

DYKDDDDK Tag Monoclonal antibody specifically detects DYKDDDDK Tag in Tag samples. It is validated for ELISA, Western Blot

Specifications

Antigen DYKDDDDK Tag
Applications ELISA, Western Blot
Classification Monoclonal
Clone 5A8E5
Conjugate Biotin
Formulation PBS with BSA and 0.02% sodium azide; pH 7.4
Gene Alias DDDDK; DDDDK epitope tag; ECS epitope tag; ECS tag; FLAG; FLAG Tag
Host Species Mouse
Immunogen A synthetic peptide (DYKDDDDK) coupled to KLH
Purification Method Protein A
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Content And Storage -20°C or -80°C if preferred
Product Type Antibody
Form Lyophilized
Isotype IgG2b κ
Target Molecule Tag
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.