Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

DYKDDDDK Tag Mouse, HRP, Clone: 5A8E5, THE™, GenScript™

Mouse Monoclonal Antibody

Supplier:  Genscript Corporation A01428100

Encompass_Preferred

Catalog No. 50-273-497


May include imposed supplier surcharges.
Only null left
Add to Cart

Description

Description

Reconstitute the lyophilized powder with deionized water (or equivalent) to an final concentration of 0.5 mg/mL. GenScript THETM DYKDDDDK Tag Antibody [HRP], mAb, Mouse is HRP conjugated THETM Anti-DYKDDDDK Monoclonal Antibody (Mouse) (A00187) purified from mice ascites by protein A affinity column. The antibody is suitable for detecting the expression level of DYKDDDDK in fusion proteins.

DYKDDDDK Tag Monoclonal antibody specifically detects DYKDDDDK Tag in Tag samples. It is validated for ELISA, Western Blot
Specifications

Specifications

DYKDDDDK Tag
Monoclonal
HRP
DDDDK; DDDDK epitope tag; ECS epitope tag; ECS tag; FLAG; FLAG Tag
A synthetic peptide (DYKDDDDK) coupled to KLH
100 μg
Primary
Antibody
IgG2b κ
ELISA, Western Blot
5A8E5
PBS with BSA and 0.01% thimerosal; pH 7.4
Mouse
Protein A
RUO
-20°C or -80°C if preferred
Lyophilized
Tag
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.