Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

DYKDDDDK Tag Mouse, iFluor 647, Clone: 5A8E5, THE™, GenScript™

Catalog No. 502120602
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-212-0602 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-212-0602 Supplier Genscript Corporation Supplier No. A01811100
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Reconstitute the lyophilized product with deionized water (or equivalent) to make antibody concentration of 0.5 mg/mL. The Flag tag is a polypeptide of with the amino acid sequence DYKDDDDK amino acid sequence that can be added to a target protein using recombinant DNA technology. It can be fused to the N-terminus or C-terminus of the protein to facilitate detection and purification. An anti-Flag tag antibody is a useful tool for the analysis of Flag-tagged proteins. THE™ DYKDDDDK Tag Antibody [iFluor 647], mAb, Mouse is THE™ DYKDDDDK Tag Antibody (A00187) conjugated with iFluor 647 under optimal conditions with an F/P ratio of 3-6. It is suitable for detecting the expression level of DYKDDDDK-tagged proteins. iFluor 647 is a bright and photostable fluorescent dye. It is an excellent alternative to Alexa Fluor 647.

DYKDDDDK Tag Monoclonal antibody specifically detects DYKDDDDK Tag in Tag samples. It is validated for Flow Cytometry, Immunocytochemistry

Specifications

Antigen DYKDDDDK Tag
Applications Flow Cytometry, Immunocytochemistry
Classification Monoclonal
Clone 5A8E5
Conjugate iFluor 647
Formulation PBS with 10mg/mL BSA and 0.02% sodium azide; pH 7.4
Gene Alias DDDDK; DDDDK epitope tag; ECS epitope tag; ECS tag; FLAG; FLAG Tag
Host Species Mouse
Immunogen A synthetic peptide DYKDDDDK coupled to KLH
Purification Method Protein A
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Content And Storage -20°C or -80°C if preferred
Product Type Antibody
Form Lyophilized
Isotype IgG2b κ
Target Molecule Tag
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.