Learn More
DYKDDDDK-tag Rabbit, Polyclonal, GenScript™
Rabbit Polyclonal Antibody
Supplier: Genscript Corporation A0017040

Description
Reconstitute the lyophilized antibody with deionized water (or equivalent) to a final concentration of 0.5 mg/mL. The DYKDDDDK peptide is a small component of the epitope which does not appear to interfere with the bioactivity or the biodistribution of the recombinant protein. It has been used extensively as a general epitope tag in expression vectors. DYKDDDDK-tag Antibody, pAb, Rabbit can be used to detect DYKDDDDK-tagged proteins.
DYKDDDDK-tag Polyclonal antibody specifically detects DYKDDDDK-tag in Tag samples. It is validated for ELISA, Flow Cytometry, Immunocytochemistry, Western BlotSpecifications
DYKDDDDK-tag | |
Polyclonal | |
PBS with 0.02% sodium azide | |
Rabbit | |
Affinity chromatography | |
RUO | |
-20°C or -80°C if preferred | |
Lyophilized | |
Tag |
ELISA, Flow Cytometry, Immunocytochemistry, Western Blot | |
Unconjugated | |
DDDDK; DDDDK epitope tag; ECS epitope tag; ECS tag; FLAG; FLAG Tag | |
A synthetic peptide DYKDDDDK (Asp-Tyr-Lys-Asp-Asp-Asp-Asp-Lys) conjugated to KLH | |
40 μg | |
Primary | |
Antibody | |
IgG |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.