Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ DYKDDDDK Tag Recombinant Rabbit Monoclonal Antibody (8H8L17), Alexa Fluor™ Plus 594, Invitrogen™

Catalog No. 502529359
Encompass_Preferred
Change view
Click to view available options
Quantity:
20 μg
50 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-252-9359 20 μg
70-162-9RP594 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 50-252-9359 Supplier Invitrogen™ Supplier No. 701629RP59420UG
Add to Cart
Edge
Add to Cart

Rabbit Recombinant Monoclonal Antibody

DYKDDDDK tag, also known as FLAG tag, is a short peptide sequence that is commonly used as a protein tag to facilitate protein purification, detection, and localization. The tag consists of eight amino acids: Asp-Tyr-Lys-Asp-Asp-Asp-Asp-Lys. It is often fused to the N- or C-terminus of a protein of interest, allowing for easy identification and isolation of the protein using anti-FLAG antibodies. The DYKDDDDK tag is widely used in molecular biology and biochemistry research, and has been employed in various applications, such as protein-protein interaction studies, protein expression analysis, and protein localization studies. The FLAG tag structure has been optimized for compatibility with the proteins it is attached to, in that it is more hydrophilic than other common epitope tags and therefore less likely to denature or inactivate proteins to which it is appended. FLAG is a registered trademark of Merck KGaA, Darmstadt, Germany.
TRUSTED_SUSTAINABILITY

Specifications

Antigen DYKDDDDK Tag
Applications Flow Cytometry, Western Blot, Immunocytochemistry
Classification Recombinant Monoclonal
Clone 8H8L17
Concentration 1.0 mg/mL
Conjugate Alexa Fluor Plus 594
Formulation Proprietary buffer with 0.008% Bromonitrodioxane, 0.008% Methylisothiazolone; pH 6.5
Gene Alias DDDDK; DDDDK epitope tag; ECS epitope tag; ECS tag; FLAG; FLAG Tag
Host Species Rabbit
Immunogen Synthetic peptide (DYKDDDDK).
Purification Method Protein A
Quantity 20 μg
Regulatory Status RUO
Primary or Secondary Primary
Target Species Tag
Content And Storage 4°C, store in dark, DO NOT FREEZE!
Product Type Antibody
Form Liquid
Isotype IgG
Target Molecule Tag
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.