Learn More
Thermo Scientific™ DyLight™ 800 NHS Ester
Description
Thermo Scientific™ Amine-Reactive DyLight Dyes are NHS ester-activated DyLight Fluors for fluorescent labeling of antibodies and other proteins to be used as molecular probes for cellular imaging and other fluorescence detection methods.
Highlights:
Brightness: high-fluorescence intensity provides outstanding sensitivity and requires less conjugate
Photostability: exceptional resistance to photobleaching enables fluorescence imaging under the most demanding conditions (e.g., structured illumination, 4pi microscopy)
Instrument compatibility: emission spectra match the principal output wavelengths of common fluorescence instrumentation
Specifications
Specifications
| Quantity | 1 mg |
| Product Type | 800 NHS Ester |
| Solubility | DMF (Dimethylformamide) |
| Content And Storage | Store desiccated at -20°C. |
| Chemical Reactivity | Amine |
| Reactive Moiety | Active Ester, Succinimidyl Ester, NHS Ester |
| Label or Dye | DyLight™ 800 |
| Label Type | DyLight Fluors |
| Product Line | DyLight™ |
| Emission | 794 |
| Show More |
For Research Use Only. Not for use in diagnostic procedures.
Your input is important to us. Please complete this form to provide feedback related to the content on this product.