Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ DYM Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA565546
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA565546 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA565546 Supplier Invitrogen™ Supplier No. PA565546
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: RMMLEIINSCL TNSLHHNPNL VYALLYKRDL FEQFRTHPSF QDIMQNIDLV ISFFSSRLLQ AGAELSVERV LEIIKQGVVA LPKDRLKKFP ELKFKYV Highest antigen sequence identity to the following orthologs - mouse 100%, rat 99%.

This gene encodes a protein which is necessary for normal skeletal development and brain function. Mutations in this gene are associated with two types of recessive osteochondrodysplasia, Dyggve-Melchior-Clausen (DMC) dysplasia and Smith-McCort (SMC) dysplasia, which involve both skeletal defects and mental retardation.
TRUSTED_SUSTAINABILITY

Specifications

Antigen DYM
Applications Immunocytochemistry
Classification Polyclonal
Concentration 0.2 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene Dym
Gene Accession No. Q7RTS9
Gene Alias 1810041M12Rik; 4933427L07Rik; C030019K18Rik; DMC; Dyggve-Melchior-Clausen syndrome protein; Dym; Dymeclin; RGD1309111; SMC
Gene Symbols Dym
Host Species Rabbit
Immunogen Recombinant Human DYM. Recombinant protein control fragment (Product #RP-106219).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 54808
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.