Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ DYM Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA597680
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA597680 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA597680 Supplier Invitrogen™ Supplier No. PA597680
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

DYM Polyclonal Antibody for Western Blot, IHC (P), ELISA

This gene encodes a protein which is necessary for normal skeletal development and brain function. Mutations in this gene are associated with two types of recessive osteochondrodysplasia, Dyggve-Melchior-Clausen (DMC) dysplasia and Smith-McCort (SMC) dysplasia, which involve both skeletal defects and mental retardation.
TRUSTED_SUSTAINABILITY

Specifications

Antigen DYM
Applications ELISA, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 3.69 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.03% ProClin 300; pH 7.4
Gene Dym
Gene Accession No. Q7RTS9, Q8CHY3
Gene Alias 1810041M12Rik; 4933427L07Rik; C030019K18Rik; DMC; Dyggve-Melchior-Clausen syndrome protein; Dym; Dymeclin; RGD1309111; SMC
Gene Symbols Dym
Host Species Rabbit
Immunogen Recombinant Human Dymeclin protein (224-348AA).
Purification Method Protein G
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 54808, 69190
Target Species Human, Mouse
Content And Storage -20°C or -80°C if preferred
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.