Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Dynabeads™ Carboxylic Acid for RNA Purification

Catalog No. p-200003584
Encompass_Preferred
Click to view available options
Quantity:
2 mL
10 mL

Dynabeads Carboxylic Acid for RNA Purification are monosized (1 μm), monodispersed, paramagnetic beads characterized by high speed to magnet and high pellet stability.

Dynabeads Carboxylic Acid for RNA Purification are monosized (1 μm), monodispersed, paramagnetic beads characterized by high speed to magnet and high pellet stability. They are intended for use with Dynabeads RNA Binding Buffer, for simple, scalable, and automation-ready purification of crude RNA produced by in vitro transcription (IVT). The combination is particularly suitable where flexibility and high purity are required, such as during the discovery phase of mRNA vaccine development and following the solid-phase IVT reaction with Dynabeads Streptavidin for In Vitro Transcription.

Features of Dynabeads Carboxylic Acid for RNA Purification include:

  • Efficiency—reuse at least six times to reduce the cost and time in a simple workflow
  • Scalability—swap between manual and automated workflows to match your required scale (micrograms to grams of mRNA) and throughput
  • Quality—ensure safe and flexible supply of magnetic beads to navigate from RNA-based drug discovery to commercial manufacturing without incurring extra time or costs from switching technology or raw material

Crude RNA from an IVT reaction step is mixed with the beads and the buffer is added. The RNA binds to the bead surface and the remaining components of the reaction mixture are removed by applying a magnet and discarding the supernatant.

The bead concentration is 10 mg/mL. They are stored in purified water and may be recycled at least six times. Scripts are available for KingFisher automated purification workflows.

The beads are offered here as a stand-alone product. The are also included in the kits below:

  • Dynabeads RNA Purification Kit (Cat. No. 65030D)
  • Dynabeads In Vitro Transcription and RNA Purification Kit (Cat. No. 65034D)

To learn more about other products and support documentation available for GMP-compliant manufacturing of mRNA vaccines and therapeutics visit: thermofisher.com/dynabeadsmrna

TRUSTED_SUSTAINABILITY

Specifications

Concentration 10 mg/mL
Color Brown
Content And Storage Dynabeads magnetic beads are supplied in purified water. Store at 2°C to 8°C.
Diameter (Metric) 1 μm
Isolation Technology Magnetic Bead
High-throughput Compatibility Yes
For Use With (Equipment) KingFisher™ Sample Purification System, DynaMag™ magnets
For Use With (Application) Synthetic RNA
Product Line Dynabeads™
Product Type Carboxylic Acid Magnetic Bead
Quantity 2 mL
Shipping Condition Ambient Temperature
Volume (Metric) 2 mL
Certifications/Compliance ISO9001 and ISO13485
Material Magnetic Polystyrene
Uniformity Monosized 1 μm (CV <5%)
Show More Show Less
Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.