Learn More
Invitrogen™ Dynabeads™ mRNA DIRECT™ Purification Kit
Description
The Dynabeads™ mRNA DIRECT™ Kit is designed for simple and rapid isolation of pure, intact polyadenylated (polyA) mRNA directly from the crude lysate of animal and plant cells and tissues. The isolated mRNA is suitable for use in all downstream applications.
Advantages of the Dynabeads mRNA DIRECT Kit:
• Fast—15 minute procedure yields pure, intact mRNA
• Highly pure mRNA isolation—best choice upstream of cDNA synthesis
• Sensitive mRNA isolation—enables cDNA synthesis and cDNA library construction from ultra-small starting samples (enables cDNA library construction from a single cell)
System overview
The isolation protocol relies on base pairing between the polyA residues at the 3' end of most mRNA, and the oligo (dT)25 residues covalently coupled to the surface of the Dynabeads. Other RNA species lacking a polyA tail will not hybridize to the beads and are readily washed away. Ribosomal RNA, DNA, proteins, and small RNA molecules (such as transfer RNA, micro RNA, and small nucleolar RNA) do not bind to the beads and are discarded. RNase inhibiting agents in the Lysis/Binding Buffer together with stringent hybridization and washing conditions ensure the isolation of pure, intact mRNA from crude samples rich in RNase, without the use of strong chaotropic agents. The mRNA purification beads specifically target and capture the mRNA transcriptome from an extremely wide variety of crude starting samples (see protocol). 1 mg of Dynabeads Oligo (dT)25 beads (200 μL) binds up to 2 μg of mRNA. A typical mammalian cell contains about 10–30 pg of total RNA, from which 1–5% is mRNA.
Specifications
Specifications
| Content And Storage | • 5 mL Dynabeads oligo (dT)25 |
| Isolation Technology | Magnetic Bead |
| Purification Time | 15 min. |
| Sample Type | Blood, Cells, FFPE and Fixed Samples, Liquid Samples (e.g. Serum), Plant Samples, RNA, Tissue, Viral Fluids, Yeast |
| Final Product Type | mRNA |
| High-throughput Compatibility | High-throughput Compatible, Manual Protocols, Automated Protocols |
| For Use With (Application) | Real-Time Quantitative PCR (qPCR), Reverse Transcriptase PCR (RT-PCR), cDNA Library Construction, Nuclease Protection Assays, Northern Blotting, Cloning |
| Product Line | DYNAL™, Dynabeads™, mRNA DIRECT™ |
| Quantity | 5 mL |
| Shipping Condition | Room Temperature |
| Show More |
Safety and Handling
For Research Use Only. Not for use in diagnostic procedures.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.