Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Dynabeads™ Protein G and Magnet Starter Pack

Catalog No. p-7110988
Encompass_Preferred
Click to view available options
Sufficient For:
100 reactions
40 reactions

The Dynabeads Protein G and Magnet Starter Pack combines Dynabeads Protein G magnetic beads for immunoprecipitation with the magnet preferred for use with 1.5 mL microcentrifuge tubes for purchasing convenience.

Space-saving design with our smallest thermal cycler footprint

  • Flexible modular design saves space on deck
  • Separate control panel module provides flexibility when integrating on the robotic deck
  • Choice of 10 cm,1 meter and 3 meter cabling options

Automated lid and cycler features

  • 384-well block
  • Automated lid is compatible with programming for easy hands-free operation with liquid handling or plate stackers
  • Heated cover slides forward to cover the plate nest,ensuring thermal contact and eliminating evaporation

For Research Use Only. Not for use in diagnostic procedures.

TRUSTED_SUSTAINABILITY

Specifications

Content And Storage The product contains:
• 2 x 1 mL Dynabeads Protein G beads
• 1 x DynaMag-2 magnet

Store Dynabeads at 2–8°C.
Shipping Condition Room Temperature
Product Line DynaMag™, Dynabeads™
Quantity 40 reactions
Type Magnet Starter Pack
Ligand Type Protein G
Sufficient For 40 reactions
WARNING: Cancer - www.P65Warnings.ca.gov

For Research Use Only. Not for use in diagnostic procedures.

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.