Learn More
Invitrogen™ Dynabeads™ Untouched™ Human CD4 T Cells Kit

Description
Includes
10mL Depletion Dynabeads and 2mL Antibody Mix with monoclonal antibodies towards human CD8, CD14, CD16 (a and b), CD19, CD36, CD56, CD123 and CD235a (Glycophorin A)
The kit depletes CD8+ T cells, B cells, NK cells, monocytes, platelets, dendritic cells, granulocytes and erythrocytes. The negatively isolated human CD4+ T-cells are left in sample and have not been in contact with Dynabeads. The bead and antibody-free CD4+ T-cells are truly untouched, and in perfect shape for any functional assay and application.
- High purity, recovery and viability of human untouched CD4+ T cells
- Easy to use and to scale up
- No columns required
- Not high throughput compatible
A negative isolation strategy isolates pure and truly untouched cells. When maximum cell purity is critical, a positive cell isolation strategy is recommended.
Order Info
Shipping Condition: Room Temperature

Specifications
Specifications
Cell Type | T Cells (CD4) |
Clonality | Monoclonal |
Content And Storage | The Kit Contains: 10 mL Depletion Dynabeads™ and 2 mL Antibody Mix with monoclonal antibodies towards human CD8, CD14, CD16 (a and b), CD19, CD36, CD56, CD123 and CD235a (Glycophorin A). Store at 2°C to 8°C. |
Product Type | Cell Isolation Kit |
Final Product Type | Cells |
High-throughput Compatibility | Not High-throughput Compatible (Manual) |
Isolation Technology | Negative Isolation |
No. of Cells | Processes ∼1 x 109 cells total |
No. of Tests | 100 Isolations |
Output Viability | >95%, >98% |
Show More |
For Research Use Only. Not for use in diagnostic procedures.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.