Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ Dynactin 1 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA583239
Description
Immunogen sequence: RELTNQQEAS VERQQQPPPE TFDFKIKFAE TKAHAKAIEM ELRQMEVAQA NRHMSLLTAF MPDSFLRPGG DHDCVLVLLL.
This gene encodes the largest subunit of dynactin, a macromolecular complex consisting of 10 subunits ranging in size from 22 to 150 kD. Dynactin binds to both microtubules and cytoplasmic dynein. Dynactin is involved in a diverse array of cellular functions, including ER-to-Golgi transport, the centripetal movement of lysosomes and endosomes, spindle formation, chromosome movement, nuclear positioning, and axonogenesis. This subunit interacts with dynein intermediate chain by its domains directly binding to dynein and binds to microtubules via a highly conserved glycine-rich cytoskeleton-associated protein (CAP-Gly) domain in its N-terminus. Alternative splicing of this gene results in multiple transcript variants encoding distinct isoforms. Mutations in this gene cause distal hereditary motor neuronopathy type VIIB (HMN7B) which is also known as distal spinal and bulbar muscular atrophy (dSBMA).
Specifications
Dynactin 1 | |
Polyclonal | |
Unconjugated | |
DCTN1 | |
150 kDa dynein-associated polypeptide; AL022633; DAP 150; DAP150; DAP-150; DCTN 1; DCTN1; DP 150; DP150; DP-150; dynactin 1; dynactin 1 (p150, glued homolog, Drosophila); dynactin 1, retrograde axonal transport; dynactin subunit 1; Dynactin1; Glued; HMN7B; OTTHUMP00000202491; p135; p150; p150 glued; p150 glued homolog; p150>glued>; p150glued; p150-glued | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
1639 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
0.10 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q14203 | |
DCTN1 | |
Recombinant protein corresponding to Human DCTN1. Recombinant protein control fragment (Product #RP-96242). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction