Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Dynactin 1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5120587
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5120587 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. PIPA5120587 Supplier Invitrogen™ Supplier No. PA5120587
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Positive test controls include: U-251MG, HeLa, HepG2, 22RV1, Jurkat, Mouse testis, Rat testis. The target is usually found in the following locations: Cytoplasm, cytoskeleton. Immunogen sequence: PGLVKDSPLL LQQISAMRLH ISQLQHENSI LKGAQMKASL ASLPPLHVAK LSHEGPGSEL PAGALYRKTS QLLETLNQLS THTHVVDITR TSPAAKSPSA QLMEQVAQLK SLSDTVEKLK DEVLKETVSQ RPGATVPTDF ATFPSSAFLR AKEEQQDDTV YMGKVTFSCA AGFGQRHRLV LTQEQLHQLH SRLIS.

This gene encodes the largest subunit of dynactin, a macromolecular complex consisting of 10 subunits ranging in size from 22 to 150 kD. Dynactin binds to both microtubules and cytoplasmic dynein. Dynactin is involved in a diverse array of cellular functions, including ER-to-Golgi transport, the centripetal movement of lysosomes and endosomes, spindle formation, chromosome movement, nuclear positioning, and axonogenesis. This subunit interacts with dynein intermediate chain by its domains directly binding to dynein and binds to microtubules via a highly conserved glycine-rich cytoskeleton-associated protein (CAP-Gly) domain in its N-terminus. Alternative splicing of this gene results in multiple transcript variants encoding distinct isoforms. Mutations in this gene cause distal hereditary motor neuronopathy type VIIB (HMN7B) which is also known as distal spinal and bulbar muscular atrophy (dSBMA).
TRUSTED_SUSTAINABILITY

Specifications

Antigen Dynactin 1
Applications ELISA, Immunoprecipitation, Western Blot
Classification Polyclonal
Concentration 3.35 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Gene DCTN1
Gene Accession No. O08788, P28023, Q14203
Gene Alias 150 kDa dynein-associated polypeptide; AL022633; DAP 150; DAP150; DAP-150; DCTN 1; DCTN1; DP 150; DP150; DP-150; dynactin 1; dynactin 1 (p150, glued homolog, Drosophila); dynactin 1, retrograde axonal transport; dynactin subunit 1; Dynactin1; Glued; HMN7B; OTTHUMP00000202491; p135; p150; p150 glued; p150 glued homolog; p150>glued>; p150glued; p150-glued
Gene Symbols DCTN1
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 945-1139 of human DCTN1 (NP_001128513.1).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 13191, 1639, 29167
Target Species Human, Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.