Learn More
Invitrogen™ Dynactin 2 Polyclonal Antibody

Sheep Polyclonal Antibody
Supplier: Invitrogen™ PA547881
Description
This antibody detects human Dynactin Subunit 2/DCTN2 in direct ELISAs and human and mouse Dynactin Subunit 2/DCTN2 in Western blots. In direct ELISAs, less than 1% cross-reactivity with recombinant human DCTN1 is observed. Reconstitute in sterile PBS to a final concentration of 0.2 mg/mL.
This gene encodes a 50-kD subunit of dynactin, a macromolecular complex consisting of 10-11 subunits ranging in size from 22 to 150 kDa. Dynactin binds to both microtubules and cytoplasmic dynein. It is involved in a diverse array of cellular functions, including ER-to-Golgi transport, the centripetal movement of lysosomes and endosomes, spindle formation, chromosome movement, nuclear positioning, and axonogenesis. This subunit is present in 4-5 copies per dynactin molecule. It contains three short alpha-helical coiled-coil domains that may mediate association with self or other dynactin subunits. It may interact directly with the largest subunit (p150) of dynactin and may affix p150 in place.
Specifications
Dynactin 2 | |
Polyclonal | |
Unconjugated | |
DCTN2 | |
2310042E05Rik; 50 kDa dynein-associated polypeptide; C130077D06Rik; C87049; DCTN2; dctn2 protein; DCTN50; DCTN-50; dynactin 2; dynactin 2 (p50); dynactin complex 50 kDa subunit; Dynactin subunit 2; dynamitin; epididymis secretory protein Li 77; GMP23-48K; Growth cone membrane protein 23-48K; HEL-S-77; hypothetical protein LOC527201; p50; p50 dynamitin; p50/dynamitin; RBP50; zgc:63867 | |
Sheep | |
Antigen affinity chromatography | |
RUO | |
10540, 69654 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Lyophilized |
Western Blot, Immunocytochemistry | |
0.2 mg/mL | |
PBS with 5% trehalose and No Preservative | |
Q13561, Q99KJ8 | |
DCTN2 | |
E. coli-derived recombinant human Dynactin Subunit 2/DCTN2 Asp277-Lys401. | |
100 μg | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.