Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ Dynactin 3 Polyclonal Antibody

Goat Polyclonal Antibody
Supplier: Invitrogen™ PA519399
Description
This antibody is predicted to react with bovine and canine based on sequence homology. This antibody is tested in Peptide ELISA: antibody detection limit dilution 16,000.
This gene encodes the smallest subunit of dynactin, a macromolecular complex consisting of 10 subunits ranging in size from 22 to 150 kD. Dynactin binds to both microtubules and cytoplasmic dynein. It is involved in a diverse array of cellular functions, including ER-to-Golgi transport, the centripetal movement of lysosomes and endosomes, spindle formation, cytokinesis, chromosome movement, nuclear positioning, and axonogenesis. This subunit, like most other dynactin subunits, exists only as a part of the dynactin complex. It is primarily an alpha-helical protein with very little coiled coil, and binds directly to the largest subunit (p150) of dynactin. Alternative splicing of this gene generates 2 transcript variants.
Specifications
| Dynactin 3 | |
| Polyclonal | |
| Unconjugated | |
| DCTN3 | |
| DCTN22; DCTN-22; DCTN3; dynactin 3; dynactin 3 (p22); dynactin complex subunit 22 kDa subunit; dynactin light chain; dynactin light chain (P24); dynactin light chain p24; Dynactin subunit 3; MGC111190; p22; p24; p24 dynactin light chain | |
| Goat | |
| Ammonium sulfate precipitation | |
| RUO | |
| 11258, 362504 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot | |
| 0.5 mg/mL | |
| TBS with 0.5% BSA and 0.02% sodium azide; pH 7.3 | |
| O75935 | |
| DCTN3 | |
| Synthetic peptide sequence (QVALGNISSKRE) corresponding to the internal amino acids of DCTN3 (aa 41-52). | |
| 100 μg | |
| Primary | |
| Human, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction