Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ Dynactin 3 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA560864
Description
Immunogen sequence: FILSQVALLE QVNALVPMLD SAHIKAVPEH AARLQRLAQI HIQQQDQCVE ITEESKALLE EYNKTTMLLS KQFVQWDELL CQLEAATQVK PA Highest antigen sequence identity to the following orthologs: Mouse - 95%, Rat - 97%.
This gene encodes the smallest subunit of dynactin, a macromolecular complex consisting of 10 subunits ranging in size from 22 to 150 kD. Dynactin binds to both microtubules and cytoplasmic dynein. It is involved in a diverse array of cellular functions, including ER-to-Golgi transport, the centripetal movement of lysosomes and endosomes, spindle formation, cytokinesis, chromosome movement, nuclear positioning, and axonogenesis. This subunit, like most other dynactin subunits, exists only as a part of the dynactin complex. It is primarily an alpha-helical protein with very little coiled coil, and binds directly to the largest subunit (p150) of dynactin. Alternative splicing of this gene generates 2 transcript variants.
Specifications
Dynactin 3 | |
Polyclonal | |
Unconjugated | |
DCTN3 | |
DCTN22; DCTN-22; DCTN3; dynactin 3; dynactin 3 (p22); dynactin complex subunit 22 kDa subunit; dynactin light chain; dynactin light chain (P24); dynactin light chain p24; Dynactin subunit 3; MGC111190; p22; p24; p24 dynactin light chain | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
11258 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin), Immunocytochemistry | |
0.2 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
O75935 | |
DCTN3 | |
Recombinant protein corresponding to Human Dynactin 3. Recombinant protein control fragment (Product #RP-92986). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction