Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
dynactin 4 Antibody - Azide and BSA Free, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP323532
Description
dynactin 4 Polyclonal antibody specifically detects dynactin 4 in Human samples. It is validated for Western Blot, Immunoprecipitation, Immunohistochemistry (Paraffin)Specifications
dynactin 4 | |
Polyclonal | |
Western Blot 1:500-1:2000, Immunoprecipitation 5-10 uL/mg of lysate, Immunohistochemistry-Paraffin 1:50-1:200 | |
dynactin 4 (p62), dynactin p62 subunit, dynactin subunit 4, Dynactin subunit p62 | |
Produced in rabbits immunized with E. coli-derived Human dynactin 4 fragmenty. | |
100 μL | |
Primary | |
Human | |
Purified |
Western Blot, Immunoprecipitation, Immunohistochemistry (Paraffin) | |
Unconjugated | |
PBS, pH 7.0 | |
Rabbit | |
Antigen and protein A Affinity-purified | |
RUO | |
51164 | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction