Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

dynactin 4 Antibody, Novus Biologicals™
SDP

Rabbit Polyclonal Antibody

$487.50

Specifications

Antigen dynactin 4
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Host Species Rabbit
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
NBP155215
SDP
View Documents
Novus Biologicals
NBP155215
100 μL
Each of 1 for $487.50
Only null left
Add to Cart
 
Description

Description

dynactin 4 Polyclonal specifically detects dynactin 4 in Human samples. It is validated for Western Blot.
Specifications

Specifications

dynactin 4
Polyclonal
Rabbit
Q9UJW0
51164
Synthetic peptides corresponding to DCTN4(dynactin 4 (p62)) The peptide sequence was selected from the N terminal of DCTN4 (NP_057305). Peptide sequence ANCFDCPGCMHTLSTRATSISTQLPDDPAKTTMKKAYYLACGFCRWTSRD.
Primary
52 kDa
Western Blot
Unconjugated
RUO
dynactin 4 (p62), dynactin p62 subunit, dynactin subunit 4, Dynactin subunit p62
DCTN4
IgG
This product is specific to Subunit or Isoform 4.
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.