Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Dynactin 4 Polyclonal Antibody
Dynactin 4 Polyclonal Antibody
Supplier: Thermo Scientific PA550392
Description
Dynactin 4 Polyclonal Antibody for IHC (P)

Specifications
Dynactin 4 | |
Polyclonal | |
Unconjugated | |
DCTN4 | |
1110001K06Rik; 4930547K17Rik; C130039E17Rik; DCTN4; Dyn4; dynactin 4; dynactin 4 (p62); dynactin 4 (p62) isoform 1; dynactin 4 (p62) isoform 2; dynactin p62 subunit; dynactin subunit 4; Dynactin subunit p62; hypothetical protein LOC550479; P62; si:ch211-174h24.2; zgc:112431 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
51164 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin) | |
2.2 mg/mL | |
PBS with 40% glycerol and 0.05% sodium azide; pH 7.4 | |
Q9UJW0 | |
DCTN4 | |
Fusion protein corresponding to residues near the C terminal of human dynactin 4 (p62). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction