Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Dynactin 4 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA530617
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA530617 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA530617 Supplier Invitrogen™ Supplier No. PA530617
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Recommended positive controls: SK-N-SH. Predicted reactivity: Mouse (95%), Rat (96%), Zebrafish (82%), Xenopus laevis (81%), Pig (91%), Rhesus Monkey (98%), Bovine (97%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.

May play an role in the regulation of Ins(1,4,5)P3 around the endoplasmic reticulum. Target protein's p62 subunit of the dynactin complex involved in the cytoplasmic dynein motor machinery.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Dynactin 4
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.99 mg/mL
Conjugate Unconjugated
Formulation 0.1M tris glycine with 20% glycerol and 0.01% thimerosal; pH 7
Gene DCTN4
Gene Accession No. Q9UJW0
Gene Alias 1110001K06Rik; 4930547K17Rik; C130039E17Rik; DCTN4; Dyn4; dynactin 4; dynactin 4 (p62); dynactin 4 (p62) isoform 1; dynactin 4 (p62) isoform 2; dynactin p62 subunit; dynactin subunit 4; Dynactin subunit p62; hypothetical protein LOC550479; P62; si:ch211-174h24.2; zgc:112431
Gene Symbols DCTN4
Host Species Rabbit
Immunogen Recombinant protein encompassing a sequence within the center region of human DCTN4. The exact sequence is proprietary.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 51164
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.