Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Dynactin Subunit 1/DCTN1 Antibody, Alexa Fluor™ 405, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP171829AF405
Description
Dynactin Subunit 1/DCTN1 Polyclonal antibody specifically detects Dynactin Subunit 1/DCTN1 in Human, Mouse samples. It is validated for Western BlotSpecifications
Dynactin Subunit 1/DCTN1 | |
Polyclonal | |
Western Blot | |
dynactin 1, dynactin 1 (p150, Glued (Drosophila) homolog), dynactin subunit 1, glued homolog, Drosophila), p135, p150 Glued (Drosophila) homolog, p150-glued | |
The immunogen for this product maps to a region between residue 1228 to 1278 of human 150 kDa Dynein-Associated Polypeptide using the numbering given in entry NP_004073.2 (GeneID 1639). | |
0.1 mL | |
Cell Cycle and Replication | |
1639 | |
Store at 4°C in the dark. | |
IgG |
Western Blot | |
Alexa Fluor 405 | |
50mM Sodium Borate | |
Rabbit | |
Immunogen affinity purified | |
RUO | |
Primary | |
Human, Mouse | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction