Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ DynaMag™-50 Magnet
GSA_VA

Catalog No. 12302d
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
12-302-D 1 Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 12-302-D Supplier Invitrogen™ Supplier No. 12302D
Only null left
Add to Cart
Add to Cart

The DynaMag-50 magnet combines a strong magnetic attraction for efficient separations with a flexible and ergonomic design that ensures comfortable working position.

The DynaMag-50 magnet combines a strong magnetic attraction for efficient separations with a flexible and ergonomic design that ensures comfortable working position. A tilting mechanism on magnet enables you to rest your elbows while pipetting. The magnet can be used for any starting sample in combination with all types of cell-isolation Dynabeads™.

  • Optimal working volume: 5—50mL
  • Magnet holds 2 x 50 mL tubes
  • Excellent control and visibility of your isolations

This magnet replaces the Dynal MPC™-50 magnet

Order Info

Shipping Condition: Room Temperature

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

TRUSTED_SUSTAINABILITY

Specifications

Content And Storage Store at room temperature.
For Use With (Equipment) DynaMag™
Shipping Condition Room Temperature
Product Line DynaMag
Quantity 1 Each
Type Cell Separation Magnet
Sample Volume 10-50 mL
WARNING: Cancer - www.P65Warnings.ca.gov

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.