Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Dynamic Diagnostics Isolation Station
missing translation for 'orderingAttributeHoverText'
Material:
Powder-coated Steel
Steel
Product Type:
Over-door Hanger
PCW Isolation Station
Description
Includes
Hardware
- Flat design allows for room where space is limited
- Optional Over-the-Door Hangers (sold separately) allow these units to be placed instantly when precautions are required
- Provides room for three glove boxes, folded gowns and facemasks
- 22G steel with rust-resistant white powder-coated finish
- Feature key holes for wall mounting for easy placement and removal of supplies
Specifications
Specifications
Product Type | PCW Isolation Station |
Color | White |
For Use With (Application) | Provides easy access to personal protection equipment in one area |
Holds | Glove boxes, face masks and gowns |
Includes | Hardware |
Material | Powder-coated Steel |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction