Learn More
Dynamic Diagnostics Monarch™ 1153™ One-Line Labeler: Labels
Specifications
Specifications
| Product Type | Label |
| Color | Orange |
| For Use With (Equipment) | Monarch 1131 Label Maker |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.