Learn More
Dynamic Diagnostics TransPak™ Pre-filled 10% Neutral Buffered Formalin (NBF)
Description
- Specimen containers pre-filled with 10% neutral buffered formalin
- Labelled with medical information for clinical use
- Contained in handy dispenser box designed with perforated tear away area on back for easy access
- Available in different volumes and packaging quantity
Specifications
Specifications
| Capacity | 80 mL |
| Color | Clear |
| Product Type | Formalin |
| Quantity | 120mL/80mL |
| Formulation | Water, Formaldehyde 37% , Selected Buffers |
| For Use With (Application) | Primary fixative and followed by secondary fixation by Alcohol, it produces a marvelous set of consistent histologic artifacts and results |
| Format | Container |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.