Learn More
Invitrogen™ Dynamin 1 Monoclonal Antibody (3G4B6)

Description
MA5-15285 targets Dynamin-1 in indirect ELISA, IHC and WB applications and shows reactivity with Human samples. The MA5-15285 immunogen is purified recombinant fragment of human Dynamin-1 expressed in E. Coli. MA5-15285 detects Dynamin-1 which has a predicted molecular weight of approximately 97kDa.
Specifications
Specifications
| Antigen | Dynamin 1 |
| Applications | ELISA, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot, Immunocytochemistry |
| Classification | Monoclonal |
| Clone | 3G4B6 |
| Concentration | Conc. Not Determined |
| Conjugate | Unconjugated |
| Formulation | Ascites with 0.03% sodium azide |
| Gene | DNM1 |
| Gene Accession No. | P21575, P39053, Q05193 |
| Gene Alias | AI838169; B-dynamin; D100; Dnm; Dnm1; Dyn1; dynamin 1; Dynamin BREDNM19; dynamin i; dynamin, brain; Dynamin1; Dynamin-1; EC 3.6.5.5; EIEE31; Ftfl; Kiaa4093; mKIAA4093; Neuronal dynamin; OTTHUMP00000215500 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.