Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ Dynamin 1 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA591865
Description
Positive Samples: K-562, A-549, SH-SY5Y, Mouse brain; Cellular Location: Cytoplasm, cytoskeleton Immunogen sequence: TGLFTPDMAF ETIVKKQVKK IREPCLKCVD MVISELISTV RQCTKKLQQY PRLREEMERI VTTHIREREG RTKEQVMLLI DIELAYMNTN H.
Dynamin is a GTPase required for endocytosis of clathrin coated vesicles from the membrane. Dynamin's involvement in endocytosis was first discovered during the characterization of the temperature sensitive parylytic shibire mutation from Drosophila melanogaster. Three closely related isoforms have been identified in mammals. Dynamin1 is expressed exclusively in neurons, dynamin2 is ubiquitously expressed and dynamin3 is expressed primarily in the testes. Each of these isoforms of dynamin also has several splice variants. This suggests that each isoform/splice variant may have separate intracellular targets or function in the budding of distinct vesicle types. Common domain features of the dynamin proteins include an amino acid terminal GTPase domain, a central pleckstrin homology domain and a highly basic proline rich carboxyl terminus with several Src homology 3 (SH3) domains. These SH3 domains target the dynamin to the clathrin coated pit. Dynamin is known to interact with at least two proteins, AP2 adaptor protein complex and amphiphysin.
Specifications
Dynamin 1 | |
Polyclonal | |
Unconjugated | |
DNM1 | |
AI838169; B-dynamin; D100; Dnm; Dnm1; Dyn1; dynamin 1; Dynamin BREDNM19; dynamin i; dynamin, brain; Dynamin1; Dynamin-1; EC 3.6.5.5; EIEE31; Ftfl; Kiaa4093; mKIAA4093; Neuronal dynamin; OTTHUMP00000215500 | |
Rabbit | |
Affinity Chromatography | |
RUO | |
13429, 140694, 1759 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
ELISA, Western Blot | |
1.2 mg/mL | |
PBS with 50% glycerol and 0.01% thimerosal; pH 7.3 | |
P21575, P39053, Q05193 | |
DNM1 | |
Recombinant fusion protein containing a sequence corresponding to amino acids 400-490 of human DNM1 (NP_0043992). | |
100 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction