Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Dynamin 2 Antibody (3F5C7) - BSA Free, Novus Biologicals™

Mouse Monoclonal Antibody
$548.00
Specifications
Antigen | Dynamin 2 |
---|---|
Clone | 3F5C7 |
Concentration | 1 mg/mL |
Dilution | Western Blot 1:100-1:2000, ELISA 1:100-1:2000 |
Applications | Western Blot, ELISA |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NBP2-61758
![]() |
Novus Biologicals
NBP261758 |
0.1 mL |
Each of 1 for $548.00
|
|
|||||
Description
Dynamin 2 Monoclonal specifically detects Dynamin 2 in Human samples. It is validated for Western Blot, ELISA.Specifications
Dynamin 2 | |
1 mg/mL | |
Western Blot, ELISA | |
Unconjugated | |
Mouse | |
CMTDI1CMT2M, CMTDIBdynamin II, DI-CMTB, DYN2EC 3.6.5.5, dynamin 2, dynamin-2, DYNII | |
DNM2 | |
IgG2a | |
Protein G purified |
3F5C7 | |
Western Blot 1:100-1:2000, ELISA 1:100-1:2000 | |
Monoclonal | |
Purified | |
Apoptosis, Membrane Trafficking and Chaperones, Neuronal Cell Markers, Neuroscience | |
1785 | |
Purified recombinant fragment of human Dynamin-2 (AA: 520-744) expressed in E. Coli. | |
Primary | |
98 kDa |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title