Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Dynamin 2 Antibody, Novus Biologicals™
SDP

Catalog No. NB300617 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB300617 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB300617 Supplier Novus Biologicals Supplier No. NB300617
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody has been used in 3 publications

Dynamin 2 Polyclonal specifically detects Dynamin 2 in Human, Mouse, Rat, Primate samples. It is validated for Western Blot.

Specifications

Antigen Dynamin 2
Applications Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Dilution Western Blot 1:1000
Formulation PBS and 1 mg/ml BSA with 0.05% Sodium Azide
Gene Alias CMTDI1CMT2M, CMTDIBdynamin II, DI-CMTB, DYN2EC 3.6.5.5, dynamin 2, dynamin-2, DYNII
Gene Symbols DNM2
Host Species Rabbit
Immunogen Synthetic Peptide: C S(760) P T P Q R R P V S S I H P P G R P P A(779)
Purification Method Affinity Purified
Quantity 100 μL
Regulatory Status RUO
Research Discipline Apoptosis, Membrane Trafficking and Chaperones, Neuronal Cell Markers, Neuroscience
Primary or Secondary Primary
Gene ID (Entrez) 1785
Test Specificity Dynamin 2 - Synapse
Target Species Human, Mouse, Rat, Primate
Content And Storage Store at -20C. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.