Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Dynamin 2 Monoclonal Antibody (3F5C7)
GREENER_CHOICE

Catalog No. FERMA548443
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
FERMA548443 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. FERMA548443 Supplier Invitrogen™ Supplier No. MA548443
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Dynamin 2 Monoclonal Antibody for Western Blot, ELISA

Dynamins represent one of the subfamilies of GTP-binding proteins. These proteins share considerable sequence similarity over the N-terminal portion of the molecule, which contains the GTPase domain. Dynamins are associated with microtubules. They have been implicated in cell processes such as endocytosis and cell motility, and in alterations of the membrane that accompany certain activities such as bone resorption by osteoclasts. Dynamins bind many proteins that bind actin and other cytoskeletal proteins. Dynamins can also self-assemble, a process that stimulates GTPase activity. Five alternatively spliced transcripts encoding different proteins have been described. Additional alternatively spliced transcripts may exist, but their full-length nature has not been determined.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Dynamin 2
Applications ELISA, Western Blot, Immunocytochemistry, Immunohistochemistry, Western Blot
Classification Monoclonal
Clone 3F5C7
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.05% sodium azide
Gene DNM2
Gene Accession No. P50570
Gene Alias b2b2159Clo; CMT2M; CMTDI1; CMTDIB; DI-CMTB; Dnm2; DYIIAAB; Dyn2; dynamin 2; dynamin II; Dynamin UDNM; Dynamin-2; DYNII; LCCS5; Udnm
Gene Symbols DNM2
Host Species Mouse
Immunogen Purified recombinant fragment of human Dynamin-2 (AA: 520-744) expressed in E. Coli.
Purification Method Protein G
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 1785
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.