Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Dynamin 3 Antibody, Novus Biologicals™
SDP

Catalog No. NB1203458 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB1203458 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB1203458 Supplier Novus Biologicals Supplier No. NB1203458

Rabbit Polyclonal Antibody has been used in 2 publications

Dynamin 3 Polyclonal specifically detects Dynamin 3 in Human, Mouse, Rat samples. It is validated for Western Blot.

Specifications

Antigen Dynamin 3
Applications Western Blot, Immunocytochemistry, Immunofluorescence
Classification Polyclonal
Concentration 1.0mg/mL
Conjugate Unconjugated
Dilution Western Blot 1:1000, Immunocytochemistry/Immunofluorescence 1:250
Formulation PBS with 0.05% Sodium Azide
Gene Alias Dyna III, dynamin 3, dynamin family member, dynamin-3, EC 3.6.5.5, MGC70433, T-dynamin, testicular
Gene Symbols DNM3
Host Species Rabbit
Immunogen Synthetic Peptide: C P(623) D K S F T E N D E N G Q A E N F(639)
Purification Method Affinity Purified
Quantity 100 μL
Regulatory Status RUO
Research Discipline Membrane Trafficking and Chaperones
Primary or Secondary Primary
Gene ID (Entrez) 26052
Test Specificity Detects a band at approximately 100 kDa.
Target Species Human, Mouse, Rat
Content And Storage Store at -20°C. Avoid freeze-thaw cycles. After opening store under sterile conditions.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.