Learn More
Invitrogen™ Dynamin 3 Polyclonal Antibody

Description
PA1-662 detects dynamin III (Dyn3) from rat, mouse, and human tissues and cells. PA1-662 has been successfully used in Western blot procedures. By Western blot, this antibody detects an ~100 kDa protein representing Dyn3 from HeLa cell lysate. PA1-662 immunizing peptide corresponds to amino acid residues 623-639 from rat Dyn3. PA1-662 immunizing peptide (Cat. # PEP-057) is available for use in neutralization and control experiments.
Specifications
Specifications
| Antigen | Dynamin 3 |
| Applications | Western Blot, Immunocytochemistry |
| Classification | Polyclonal |
| Concentration | Conc. Not Determined |
| Conjugate | Unconjugated |
| Formulation | PBS with 1 mg/mL BSA and 0.05% sodium azide |
| Gene | Dnm3 |
| Gene Accession No. | Q08877, Q8BZ98, Q9UQ16 |
| Gene Alias | 9630020E24Rik; AW061159; B230343F03Rik; C530045C17; DNM3; Dyn3; Dyna III; dynamin 3; dynamin family member; Dynamin III; dynamin, testicular; dynamin-2; Dynamin-3; KIAA0820; mKIAA0820; T-dynamin; testicular dynamin |
| Gene Symbols | Dnm3 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.