Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ DyNAmo Flash SYBR Green qPCR Kit

Catalog No. F415XL Shop All Thermo Scientific Products
Change view
Click to view available options
No. of Reactions:
2500 Reactions
500 reactions
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. No. of Reactions
F415XL 2500 Reactions
F415L 500 reactions
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. F415XL Supplier Thermo Scientific™ Supplier No. F415XL
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Developed for fast real-time qPCR, the kit provides qPCR results faster than most SYBR Green kits without compromising the qPCR performance.

A DNA-binding domain attached to the polymerase in this kit improves the physical stability of the polymerase-DNA complex, thus enhancing the processivity and robustness of the amplification. The high amplification efficiency of DyNAmo Flash SYBR Green qPCR Kit gives reliable quantification and early Cq values. Due to the high signal intensity, DyNAmo Flash SYBR Green qPCR provides an excellent signal-to-noise ratio.

  • Extremely fast protocols: Combined annealing and extension step of only 15 sec.
  • Specific and sensitive detection of a wide range of template concentrations
  • Included dUTP allows the use of UNG for prevention of carry-over contamination
  • Convenient 2X master mix and optimized protocol
TRUSTED_SUSTAINABILITY

Specifications

Concentration 2X (qPCR Mix), 50X (Dye)
Content And Storage Contains:
• 1 x 1 mL DyNAmo Flash SYBR Green qPCR Mix (2X)
• 1 x 1.25 mL 50 x ROX dye, 25μM

Store at -20°C
Detection Method SYBR
Format Tube
GC-Rich PCR Performance High
PCR Method qPCR
Polymerase Tbr DNA polymerase
Reaction Speed Fast
For Use With (Equipment) 7000 System, 7300 System, 7500 Fast System, 7500 System, 7700 System, 7900HT Fast System, 7900HT System, BioRad CFX96™, QuantStudio™, ViiA™ 7 System, Bio-Rad CFX384 Touch™, Roche LightCycler 480, 5700 Sequence Detection System, StepOnePlus™ System, Eppendorf MasterCycler RealPlex, Corbett RotorGene, StepOne™ System, Stratagene Mx4000, Stratagene Mx3005P, Stratagene Mx3000P, MJ Opticon, MJ Chromo4, Cepheid SmartCycler
Kit Type DyNAmo Flash SYBR Green qPCR Kit
Product Line DyNAmo Flash
Product Type Real Time PCR SYBR Master Mix
Quantity 2,500 reactions
Sufficient For 2500 Reactions of 20 μL, 1000 reactions of 50 μL
For Use With (Application) Gene Expression
No. of Reactions 2500 Reactions
Sample Type DNA, dsDNA
Volume 25 x 1 mL
Show More Show Less
Do I need to use Fast plastics to run a Fast qPCR reaction?

Specialized Fast PCR plates are not required if you are using a standard qPCR machine with a fast protocol. However, Fast Block PCR Plates (AB-1900) can be purchased which are compatible with the thermal blocks on fast qPCR thermal cyclers.

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.