Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ Dynapyrazole A
GREENER_CHOICESDP

Catalog No. 689625 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
5 mg
25 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
68-962-5 25 mg
68-965 5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 68-962-5 Supplier Tocris Bioscience™ Supplier No. 6896/25
Add to Cart
Add to Cart
This item is not returnable. View return policy

Hedgehog signaling inhibitor; dynein inhibitor

Dynapyrazole A is a dynein inhibitor. Inhibits dynein 1 and dynein 2-dependent microtubule gliding (IC50 values are 2.3 and 2.6 μM, respectively). Inhibits hedgehog signaling in vitro (IC50 = 1.9 μM). Displays reversible inhibition of retrograde and anterograde intraflagellar transport in Chlamydomonas; displays stronger inhibition of retrograde transport.

Specifications

Chemical Name or Material 2-[1-(4-Chlorophenyl)cyclopropyl]-4,5-dihydro-7-iodo-5-oxopyrazolo[1,5-a]quinazoline-3-carbonitrile
CAS 2226517-75-3
Quantity 25 mg
Target Hedgehog Signaling Inhibitors
Molecular Formula C20H12ClIN4O
Formula Weight 3333
Purity 98%
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.