Learn More
Dynatech Precision Sampling Heavy Duty Purge and Trap Syringe from Restek
SDP
Catalog No. 06711462 Shop All Restek Corporation Products
Encompass_Preferred
Change view
Click to view available options
Volume (Metric):
2 mL
5 mL
10 mL
Includes:
Not Applicable
Sample lok valve
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Volume (Metric) Includes
06-711-462 10 mL Sample lok valve
06-711-463 5 mL Sample lok valve
06-711-464 10 mL Not Applicable
06-711-461 5 mL Not Applicable
06-711-460 2 mL Not Applicable
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. 06-711-462 Supplier Restek Corporation Supplier No. 21207
Add to Cart
Add to Cart
Heavy-duty glass barrel with metal front and rear flanges.
  • PTFE luer lock tip
  • Can fill and empty sparge tubes
  • Sample-Lok valve accepts standard luer lock needles
  • Suitable for purge and trap applications
Specifications
Material PTFE (Tip)
For Use With (Application) For purge and trap applications
Includes Sample lok valve
Product Type Syringe
Quantity Ea.
Volume (Metric) 10 mL
Connection Type Luer Lock