Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ DYNC1H1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5117280
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5117280 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5117280 Supplier Invitrogen™ Supplier No. PA5117280
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Centrifuge briefly prior to opening.

DYNC1H1 (Dynein Cytoplasmic 1 Heavy Chain 1) is a gene that encodes a protein belonging to the dynein family, which are motor proteins critical for various cellular processes. Dyneins are responsible for retrograde intracellular transport along microtubules, meaning they move cargo from the periphery of the cell towards the nucleus. The DYNC1H1 protein is a heavy chain component of cytoplasmic dynein 1, which plays a key role in the movement of organelles, vesicles, and other cellular components. Mutations in DYNC1H1 have been associated with several neurological disorders, including spinal muscular atrophy with lower extremity predominance (SMA-LED) and Charcot-Marie-Tooth disease type 2O (CMT2O). Research into DYNC1H1 continues to uncover its involvement in cellular mechanics and its implications in various genetic conditions.
TRUSTED_SUSTAINABILITY

Specifications

Antigen DYNC1H1
Applications Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.89 mg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 20% glycerol and 0.01% thimerosal; pH 7
Gene dync1h1
Gene Accession No. Q14204
Gene Alias 9930018I23Rik; AI894280; CMT2O; cytoplasmic dynein 1 heavy chain 1; Cytoplasmic dynein heavy chain 1; cytoplasmic dynein heavy chain polypeptide 1; Dhc1; DHC1a; DNCH1; Dnchc1; DNCL; Dnec1; DNECL; DYHC; DYNC1H1; dynein cytoplasmic 1 heavy chain 1; dynein heavy chain, cytosolic; dynein heavy chain, retrograde transport; dynein, cytoplasmic 1, heavy chain 1; dynein, cytoplasmic, heavy chain 1; dynein, cytoplasmic, heavy polypeptide 1; fk70a07; HL-3; KIAA0325; legs at odd angle; Loa; MAP 1C; MAP1C; mKIAA0325; p22; SMALED1; sprawling; Swl; wu:fk70a07
Gene Symbols dync1h1
Host Species Rabbit
Immunogen Recombinant protein corresponding to human DYNC1H1.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 1778
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.