Learn More
Invitrogen™ DYNC1H1 Polyclonal Antibody

Description
DYNC1H1 Polyclonal Antibody for Western Blot, IHC (P)
Specifications
Specifications
| Antigen | DYNC1H1 |
| Applications | Immunohistochemistry (Paraffin), Western Blot, Western Blot |
| Classification | Polyclonal |
| Concentration | 0.5 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 0.09% sodium azide; pH 7.4 |
| Gene | dync1h1 |
| Gene Accession No. | Q14204, Q9JHU4 |
| Gene Alias | 9930018I23Rik; AI894280; CMT2O; cytoplasmic dynein 1 heavy chain 1; Cytoplasmic dynein heavy chain 1; cytoplasmic dynein heavy chain polypeptide 1; Dhc1; DHC1a; DNCH1; Dnchc1; DNCL; Dnec1; DNECL; DYHC; DYNC1H1; dynein cytoplasmic 1 heavy chain 1; dynein heavy chain, cytosolic; dynein heavy chain, retrograde transport; dynein, cytoplasmic 1, heavy chain 1; dynein, cytoplasmic, heavy chain 1; dynein, cytoplasmic, heavy polypeptide 1; fk70a07; HL-3; KIAA0325; legs at odd angle; Loa; MAP 1C; MAP1C; mKIAA0325; p22; SMALED1; sprawling; Swl; wu:fk70a07 |
| Gene Symbols | dync1h1 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.