Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
DYNC1H1 Rabbit anti-Human, Rat, Polyclonal, Invitrogen™
Rabbit Polyclonal Antibody
Supplier: Thermo Scientific PA549373
Description
Predicted to react with mouse based on sequence homology.
Cytoplasmic dynein 1 acts as a motor for the intracellular retrograde motility of vesicles and organelles along microtubules. Dynein has ATPase activity; the force-producing power stroke is thought to occur on release of ADP.
Specifications
| DYNC1H1 | |
| Polyclonal | |
| Unconjugated | |
| DYNC1H1 | |
| Cytoplasmic dynein 1 heavy chain 1, Cytoplasmic dynein heavy chain 1, Dynein heavy chain, cytosolic, DYNC1H1, DHC1, DNCH1, DNCL, DNECL, DYHC, KIAA0325 | |
| Rabbit | |
| Protein A, Antigen Affinity Chromatography | |
| RUO | |
| 1778, 29489 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Western Blot | |
| 0.29 mg/mL | |
| PBS with 0.09% sodium azide | |
| Q14204, P38650 | |
| DYNC1H1 | |
| KLH conjugated synthetic peptide between 4224-4257 amino acids from the C-terminal region of human DYNC1H1. | |
| 400 μL | |
| Primary | |
| Human, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction