Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ DYNC1I1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA597681
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA597681 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA597681 Supplier Invitrogen™ Supplier No. PA597681
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

DYNC1I1 Polyclonal Antibody for Western Blot, IHC (P), ELISA

Acts as one of several non-catalytic accessory components of the cytoplasmic dynein 1 complex that are thought to be involved in linking dynein to cargos and to adapter proteins that regulate dynein function. Cytoplasmic dynein 1 acts as a motor for the intracellular retrograde motility of vesicles and organelles along microtubules. The intermediate chains mediate the binding of dynein to dynactin via its 150 kDa component (p150-glued) DCNT1. May play a role in mediating the interaction of cytoplasmic dynein with membranous organelles and kinetochores.
TRUSTED_SUSTAINABILITY

Specifications

Antigen DYNC1I1
Applications ELISA, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.23 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Gene DYNC1I1
Gene Accession No. O14576, O88485
Gene Alias Cytoplasmic dynein 1 intermediate chain 1; cytoplasmic dynein intermediate chain 1; cytoplasmic dynein intermediate chain 1 isoform 1.6; DH IC-1; DHIC-1; DIC; Dnci 1; DNCI1; Dncic1; Dync1i1; dynein cytoplasmic 1 intermediate chain 1; dynein intermediate chain 1, cytosolic; Dynein intermediate chain 1, cytosolic (DH IC-1) (Cytoplasmic dynein intermediate chain 1); dynein, cytoplasmic 1, intermediate chain 1; dynein, cytoplasmic, intermediate chain 1; dynein, cytoplasmic, intermediate polypeptide 1; IC74
Gene Symbols DYNC1I1
Host Species Rabbit
Immunogen Recombinant Human Cytoplasmic dynein 1 intermediate chain 1 protein (1-220AA).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 13426, 1780
Target Species Human, Mouse
Content And Storage -20°C or -80°C if preferred
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.