Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ DYNC1I2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA562892
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA562892 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA562892 Supplier Invitrogen™ Supplier No. PA562892
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: EDDDVVAPKP PIEPEEEKTL KKDEENDSKA PPHELTE Highest antigen sequence identity to the following orthologs: Mouse - 89%, Rat - 89%.

Dyneins are multi-subunit, high molecular weight ATPases that interact with microtubules to generate force by converting the chemical energy of ATP into the mechanical energy of movement. Cytoplasmic or axonemal Dynein heavy, intermediate, light and light-intermediate chains are all components of minus end-directed motors; the complex that transports cellular cargos towards the central region of the cell. Axonemal Dynein motors contain one to three non-identical heavy chains and cause a sliding of microtubules in the axonemes of cilia and flagella in a mechanism necessary for cilia to beat and propel the cell. Cytoplasmic Dynein is an approximately 12 subunit complex of two heavy chains, two intermediate chains to anchor Dynein to its cargo, four smaller intermediate chains and several light chains. It performs functions necessary for cell survival such as organelle transport and centrosome assembly. The carboxy-terminus of Dynein is important for microtubule-dependent motility and is highly conserved, while the amino-terminal regions are more variable. Several proteins regulate Dynein activity, including Dynactin, LIS1 and nudel (nudE-like).
TRUSTED_SUSTAINABILITY

Specifications

Antigen DYNC1I2
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.2 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene DYNC1I2
Gene Accession No. Q13409
Gene Alias 3110079H08Rik; AW554389; Cytoplasmic dynein 1 intermediate chain 2; cytoplasmic dynein intermediate chain 2; cytoplasmic dynein intermediate chain 2 isoform 2.1; cytoplasmic dynein intermediate chain 2 isoform 2.2; cytoplasmic dynein intermediate chain 2 isoform 2.5; DH IC-2; DIC74; DNCI2; DNCIC2; Dync1i2; dynein cytoplasmic 1 intermediate chain 2; dynein intermediate chain 2, cytosolic; dynein, cytoplasmic 1, intermediate chain 2; dynein, cytoplasmic, intermediate chain 2; dynein, cytoplasmic, intermediate polypeptide 2; IC2; oncofetal protein (OFP); testis tissue sperm-binding protein Li 66n
Gene Symbols DYNC1I2
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human DYNC1I2. Recombinant protein control fragment (Product #RP-103172).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 1781
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.