Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
DYNC1LI1 Antibody (OTI2E8), DyLight 680, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP271904FR
Description
DYNC1LI1 Monoclonal specifically detects DYNC1LI1 in Human, Mouse, Rat samples. It is validated for Western Blot, Flow Cytometry, Immunocytochemistry/Immunofluorescence.Specifications
DYNC1LI1 | |
Monoclonal | |
DyLight 680 | |
PBS with 0.05% Sodium Azide | |
DYNC1LI1 | |
Full length recombinant protein of human DYNC1LI1 (NP_057225) produced in HEK293T cells. | |
0.1 mL | |
51143.0 | |
Store at 4°C in the dark. |
Western Blot, Flow Cytometry, Immunocytochemistry, Immunofluorescence | |
OTI2E8 | |
Western Blot, Flow Cytometry, Immunocytochemistry/Immunofluorescence | |
cytoplasmic dynein 1 light intermediate chain 1, DNCLI1, dynein, cytoplasmic 1, light intermediate chain 1, LIC1 | |
Mouse | |
Affinity Purified | |
Primary | |
Human, Mouse, Rat | |
IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction