Learn More
Invitrogen™ DYNC1LI1 Monoclonal Antibody (OTI1D1)

Description
DYNC1LI1 Monoclonal Antibody for Western Blot, ICC/IF, Flow
Specifications
Specifications
| Antigen | DYNC1LI1 |
| Applications | Flow Cytometry, Western Blot, Immunocytochemistry |
| Classification | Monoclonal |
| Clone | OTI1D1 |
| Concentration | 0.51 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 1% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3 |
| Gene | DYNC1LI1 |
| Gene Accession No. | Q9Y6G9 |
| Gene Alias | 1110053F02Rik; Cytoplasmic dynein 1 light intermediate chain 1; DLC-A; DNCLI1; Dnclic1; Dync1li1; dynein cytoplasmic 1 light intermediate chain 1; dynein light chain A; Dynein light intermediate chain 1, cytosolic; dynein, cytoplasmic, light intermediate chain 1; dynein, cytoplasmic, light intermediate polypeptide 1; LIC1; LIC-1 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.