Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

DYNC1LI1 Monoclonal Antibody (OTI2E8), TrueMAB™, OriGene
SDP

Catalog No. 502542227
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
30 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-254-2227 30 μL
50-167-2650 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 50-254-2227 Supplier OriGene Supplier No. TA501509S
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

DYNC1LI1 Monoclonal antibody specifically detects DYNC1LI1 in Human samples. It is validated for Flow Cytometry, Immunocytochemistry, Western Blot

Specifications

Antigen DYNC1LI1
Applications Flow Cytometry, Immunocytochemistry, Western Blot
Classification Monoclonal
Clone OTI2E8
Concentration 0.34 mg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.02% sodium azide
Gene DYNC1LI1
Gene Accession No. Q9Y6G9
Gene Alias 1110053F02Rik; Cytoplasmic dynein 1 light intermediate chain 1; DLC-A; DNCLI1; Dnclic1; Dync1li1; dynein cytoplasmic 1 light intermediate chain 1; dynein light chain A; Dynein light intermediate chain 1, cytosolic; dynein, cytoplasmic, light intermediate chain 1; dynein, cytoplasmic, light intermediate polypeptide 1; LIC1; LIC-1
Host Species Mouse
Immunogen Full length human recombinant protein of human DYNC1LI1 produced in HEK293T cell.
Purification Method Affinity chromatography
Quantity 30 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 51143
Target Species Human
Content And Storage -20° C, Avoid Freeze/Thaw Cycles
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.