Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ DYNC1LI2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5100491
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5100491 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5100491 Supplier Invitrogen™ Supplier No. PA5100491
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Antibody detects endogenous levels of total DYNC1LI2.

Cytoplasmic dynein is a microtubule-associated motorprotein.
TRUSTED_SUSTAINABILITY

Specifications

Antigen DYNC1LI2
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene DYNC1LI2
Gene Accession No. O43237, Q62698, Q6PDL0
Gene Alias AA409702; C920003I06; cytoplasmic dynein 1 light intermediate chain 2; Dncli2; Dnclic2; Dync1li2; dynein cytoplasmic 1 light intermediate chain 2; dynein light intermediate chain 2, cytosolic; dynein, cytoplasmic 1 light intermediate chain 2; dynein, cytoplasmic 1, light intermediate chain 2; dynein, cytoplasmic, light intermediate chain 2; dynein, cytoplasmic, light intermediate polypeptide 2; fc08b04; hypothetical protein LOC550362; LIC2; LIC-2; LIC-2 dynein light intermediate chain 53/55; LIC53/55; wu:fc08b04; zgc:112480
Gene Symbols DYNC1LI2
Host Species Rabbit
Immunogen A synthesized peptide derived from human DYNC1LI2(Accession O43237), corresponding to amino acid residues E172-N222.
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 1783, 234663, 81655
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.