Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ DYNC1LI2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA525392
Change view
Click to view available options
Quantity:
400 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA525392 400 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA525392 Supplier Invitrogen™ Supplier No. PA525392
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

This antibody is predicted to react with non-human primate based on sequence homology.

Cytoplasmic dynein is a microtubule-associated motorprotein.
TRUSTED_SUSTAINABILITY

Specifications

Antigen DYNC1LI2
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.09% sodium azide; pH 7.4
Gene DYNC1LI2
Gene Accession No. O43237
Gene Alias AA409702; C920003I06; cytoplasmic dynein 1 light intermediate chain 2; Dncli2; Dnclic2; Dync1li2; dynein cytoplasmic 1 light intermediate chain 2; dynein light intermediate chain 2, cytosolic; dynein, cytoplasmic 1 light intermediate chain 2; dynein, cytoplasmic 1, light intermediate chain 2; dynein, cytoplasmic, light intermediate chain 2; dynein, cytoplasmic, light intermediate polypeptide 2; fc08b04; hypothetical protein LOC550362; LIC2; LIC-2; LIC-2 dynein light intermediate chain 53/55; LIC53/55; wu:fc08b04; zgc:112480
Gene Symbols DYNC1LI2
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide between 152-180 amino acids from the central region of human DYNC1LI2.
Purification Method Antigen affinity chromatography, Protein A
Quantity 400 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 1783
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.