Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ DYNC1LI2 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA545861
Description
Peptide sequence: KLPSGKNILV FGEDGSGKTT LMTKLQGAEH GKKGRGLEYL YLSVHDEDRD Sequence homology: Cow: 100%; Dog: 100%; Guinea Pig: 100%; Horse: 100%; Human: 100%; Mouse: 100%; Rabbit: 100%; Rat: 100%; Zebrafish: 93%.
Cytoplasmic dynein is a microtubule-associated motorprotein.
Specifications
DYNC1LI2 | |
Polyclonal | |
Unconjugated | |
DYNC1LI2 | |
AA409702; C920003I06; cytoplasmic dynein 1 light intermediate chain 2; Dncli2; Dnclic2; Dync1li2; dynein cytoplasmic 1 light intermediate chain 2; dynein light intermediate chain 2, cytosolic; dynein, cytoplasmic 1 light intermediate chain 2; dynein, cytoplasmic 1, light intermediate chain 2; dynein, cytoplasmic, light intermediate chain 2; dynein, cytoplasmic, light intermediate polypeptide 2; fc08b04; hypothetical protein LOC550362; LIC2; LIC-2; LIC-2 dynein light intermediate chain 53/55; LIC53/55; wu:fc08b04; zgc:112480 | |
Rabbit | |
Affinity Chromatography | |
RUO | |
1783 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
Western Blot | |
0.5 mg/mL | |
PBS with 2% sucrose and 0.09% sodium azide | |
O43237 | |
DYNC1LI2 | |
Synthetic peptide directed towards the N-terminal of human DYNC1LI2 (aa 50-99). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction