Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ DYNC2H1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5113133
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5113133 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5113133 Supplier Invitrogen™ Supplier No. PA5113133
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

DYNC2H1 Polyclonal Antibody for Western Blot, ICC/IF, IHC (P), ELISA

TRUSTED_SUSTAINABILITY

Specifications

Antigen DYNC2H1
Applications ELISA, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 3.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.03% ProClin 300; pH 7.4
Gene DYNC2H1
Gene Accession No. Q8NCM8
Gene Alias 4432416O06Rik; AI448217; ATD3; b2b414Clo; Cytoplasmic dynein 2 heavy chain; Cytoplasmic dynein 2 heavy chain 1; cytoplasmic dynein heavy chain 2; D030010H02Rik; D330044F14Rik; DHC1b; DHC2; Dlp4; Dnch2; Dnchc2; DYH1B; Dync2h1; dynein cytoplasmic 2 heavy chain 1; dynein cytoplasmic heavy chain 2; Dynein heavy chain 11; dynein heavy chain 2; dynein heavy chain isotype 1B; dynein heavy chain, isotype 1B; dynein, cytoplasmic 2, heavy chain 1; dynein, cytoplasmic, heavy chain 2; dynein, cytoplasmic, heavy polypeptide 2; dynein-like protein 4; hDHC11; KIAA1997; mDHC11; mKIAA1997; SRPS2B; SRTD3
Gene Symbols DYNC2H1
Host Species Rabbit
Immunogen Recombinant Human Cytoplasmic dynein 2 heavy chain 1 protein (1928-2065AA).
Purification Method Protein G
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 79659
Target Species Human
Content And Storage -20°C or -80°C if preferred
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.