Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ DYNC2H1 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA558627
Description
Immunogen sequence: DVFNQRNKKS IFPYSVSLPQ SCSILDYRAV IEKIPEDDKP SFFGLPANIA RSSQRMISSQ VISQLRILGR SITAGSKFDR EIWSNELSPV LNLWKKLN Highest antigen sequence identity to the following orthologs: Mouse - 94%, Rat - 93%.

Specifications
DYNC2H1 | |
Polyclonal | |
Unconjugated | |
DYNC2H1 | |
4432416O06Rik; AI448217; ATD3; b2b414Clo; Cytoplasmic dynein 2 heavy chain; Cytoplasmic dynein 2 heavy chain 1; cytoplasmic dynein heavy chain 2; D030010H02Rik; D330044F14Rik; DHC1b; DHC2; Dlp4; Dnch2; Dnchc2; DYH1B; Dync2h1; dynein cytoplasmic 2 heavy chain 1; dynein cytoplasmic heavy chain 2; Dynein heavy chain 11; dynein heavy chain 2; dynein heavy chain isotype 1B; dynein heavy chain, isotype 1B; dynein, cytoplasmic 2, heavy chain 1; dynein, cytoplasmic, heavy chain 2; dynein, cytoplasmic, heavy polypeptide 2; dynein-like protein 4; hDHC11; KIAA1997; mDHC11; mKIAA1997; SRPS2B; SRTD3 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
79659 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin), Immunocytochemistry | |
0.2 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q8NCM8 | |
DYNC2H1 | |
Recombinant protein corresponding to Human DYNC2H1. Recombinant protein control fragment (Product #RP-97533). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction